Novus Biologicals
Manufacturer Code:NBP297888DL594
Catalog # NB049015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SGLT1/SLC5A1 fragment. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D22S675 High affinity sodium-glucose cotransporter Na+/glucose cotransporter 1 NAGTsodium/glucose cotransporter 1 SGLT1Na(+)/glucose cotransporter 1 solute carrier family 5 (sodium/glucose cotransporter) member 1 Solute carrier family 5 member 1; High affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 1; Na+/glucose cotransporter 1; Sodium/glucose cotransporter 1; solute carrier family 5 (sodium/glucose cotransporter), member 1; Solute carrier family 5 member 1
Gene Aliases: D22S675; NAGT; SGLT1; SLC5A1
UniProt ID: (Human) P13866
Entrez Gene ID: (Human) 6523
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.