Invitrogen
Manufacturer Code:PA537937
Catalog # PIPA537937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KDTIEIDAEAPQKEK from the internal region of rat Slc5a1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: High affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 1; Sodium/glucose cotransporter 1; Solute carrier family 5 member 1; solute carrier family 5 sodium/glucose cotransporter member 1 MGC93553 Na +/glucose cotransporter 1 Solute carrier family 5 member alpha 1 Na+/glucose cotransporter high affinity sodium-glucose cotransporter; solute carrier family 5, member 1; Solute carrier family 5, member alpha 1 (Na+/glucose cotransporter)
Gene Aliases: SGLT1; Slc5a1
UniProt ID: (Rat) P53790
Entrez Gene ID: (Rat) 25552
Molecular Function:
carbohydrate transporter
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.