Invitrogen
Manufacturer Code:PA534208
Catalog # PIPA534208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 14 amino acid peptide from internal region of human SLC5A1 / SGLT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 1; Na+/glucose cotransporter 1; SLC5A1 D22S675 HSGLT1 Na(+)/glucose cotransporter 1 Na+/glucose cotransporter 1 SGLT1 NAGT Sodium-glucose transporter 1 Sodium/glucose cotransporter 1; Sodium/glucose cotransporter 1; solute carrier family 5 (sodium/glucose cotransporter), member 1; Solute carrier family 5 member 1
Gene Aliases: D22S675; NAGT; SGLT1; SLC5A1
UniProt ID: (Human) P13866
Entrez Gene ID: (Human) 6523
Molecular Function: carbohydrate transporter cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.