Abnova
Manufacturer Code:PAB31641
Catalog # 89-999-257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 10-90 of human SGK1 (phospho S78). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: serine/threonine protein kinase SGK; Serine/threonine-protein kinase Sgk1; Serum/glucocorticoid-regulated kinase 1; SGK; Sgk1 variant i3
Gene Aliases: SGK; SGK1
UniProt ID: (Human) O00141
Entrez Gene ID: (Human) 6446
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.