Invitrogen
Manufacturer Code:PA530720
Catalog # PIPA530720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 232 of Human SGCE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dystonia 11, myoclonic; DYT11 ESG dystonia 11 myoclonic epsilon-SG epsilon-sarcoglycan; Epsilon-sarcoglycan; Epsilon-SG; sarcoglycan, epsilon
Gene Aliases: DYT11; ESG; SGCE; UNQ433/PRO840
UniProt ID: (Human) O43556
Entrez Gene ID: (Human) 8910
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.