Novus Biologicals
Manufacturer Code:NBP15979420UL
Catalog # NBP15979420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SGCE(sarcoglycan epsilon) The peptide sequence was selected from the N terminal of SGCE. Peptide sequence TVYSIFSKVHSDRNVYPSAGVLFVHVLEREYFKGEFPPYPKPGEISNDPI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dystonia 11 myoclonic DYT11 epsilon-sarcoglycan Epsilon-SG ESG sarcoglycan epsilon; dystonia 11, myoclonic; Epsilon-sarcoglycan; Epsilon-SG; sarcoglycan, epsilon
Gene Aliases: DYT11; ESG; SGCE; UNQ433/PRO840
UniProt ID: (Human) O43556
Entrez Gene ID: (Human) 8910
Molecular Function: calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.