Proteintech
Manufacturer Code:124071AP150UL
Catalog # 50-172-7961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SGCE Fusion Protein Ag3118 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dystonia 11, myoclonic; DYT11 Epsilon sarcoglycan Epsilon SG ESG sarcoglycan epsilon SGCE epsilon-sarcoglycan; Epsilon-sarcoglycan; epsilon-sarcoglycan {ECO:0000312|EMBL:AAO37579.1}; Epsilon-SG; epsilon-SG {ECO:0000250|UniProtKB:O43556}; sarcoglycan, epsilon; sgce {ECO:0000312|EMBL:AAO37579.1, ECO:0000312|RGD:1303201}
Gene Aliases: DYT11; e-SG; ESG; SGCE; UNQ433/PRO840
UniProt ID: (Human) O43556, (Mouse) O70258, (Rat) Q6YAT4
Entrez Gene ID: (Human) 8910, (Mouse) 20392, (Rat) 432360
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.