Invitrogen
Manufacturer Code:PA570804
Catalog # PIPA570804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat SFXN5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: sideroflexin 5 SFXN5 Park3 (&quest); Sideroflexin-5; Tricarboxylate carrier BBG-TCC
Gene Aliases: BBG-TCC; C230001H08Rik; SFXN5
UniProt ID: (Human) Q8TD22, (Mouse) Q925N0, (Rat) Q8CFD0
Entrez Gene ID: (Human) 94097, (Rabbit) 100358331, (Horse) 100050079, (Bovine) 528468, (Dog) 612350, (Mouse) 94282, (Guinea pig) 100718854, (Rat) 261737
Molecular Function:
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.