Abnova
Manufacturer Code:H00081855B01P
Catalog # 89-017-176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SFXN3 (NP_112233.2 1 a.a. ~ 325 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SFXN3 protein.
For Research Use Only
Protein Aliases: BA108L7.2; SFX3; Sideroflexin-3
Gene Aliases: BA108L7.2; SFX3; SFXN3
UniProt ID: (Human) Q9BWM7
Entrez Gene ID: (Human) 81855
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.