Proteintech
Manufacturer Code:151561AP150UL
Catalog # 50-173-0026
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SFXN3 Fusion Protein Ag7203 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SFX3 SFXN3 sideroflexin 3; Sideroflexin-3; tricarboxylate carrier; tricarboxylate carrier-like protein
Gene Aliases: BA108L7.2; SFX3; SFXN3; TCC
UniProt ID: (Human) Q9BWM7, (Rat) Q9JHY2, (Mouse) Q91V61
Entrez Gene ID: (Human) 81855, (Rat) 65042, (Mouse) 94280
Molecular Function:
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.