Proteintech
Manufacturer Code:122961AP150UL
Catalog # 50-172-7869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SFXN1 Fusion Protein Ag2985 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: flexed tail; flexed-tail; SFXN1 sideroflexin 1 TCC Tricarboxylate carrier protein; Sideroflexin-1; tricarboxylate carrier protein
Gene Aliases: 2810002O05Rik; A930015P12Rik; f; SFXN1; TCC
UniProt ID: (Human) Q9H9B4, (Mouse) Q99JR1
Entrez Gene ID: (Human) 94081, (Mouse) 14057
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.