Invitrogen
Manufacturer Code:MA140247
Catalog # 01-676-580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | VIF9 |
Immunogen | Rat SP-D. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Collectin-7; CP4; Lung surfactant protein D; PSP-D; pulmonary surfactant apoprotein; pulmonary surfactant protein D; Pulmonary surfactant-associated protein D; surfactant associated protein D; surfactant protein D SFTPD; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4
Gene Aliases: COLEC7; PSP-D; PSPD; SFTP4; SFTPD; SP-D; SPD
UniProt ID: (Human) P35247, (Rat) P35248
Entrez Gene ID: (Human) 6441, (Rat) 25350
Molecular Function:
defense/immunity protein
surfactant
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.