Invitrogen
Manufacturer Code:MA531243
Catalog # PIMA531243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 19 |
Immunogen | Recombit Mouse SFTPD protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Lung surfactant protein D; PSP-D; Pulmonary surfactant-associated protein D; Pulmonary surfactant-associated protein D SP-D PSP-D COLEC7 Collectin7 Pulmonary surfactant apoprotein SFTP4 SFTPD Surfactant associated prote
Gene Aliases: AI573415; Sftp4; Sftpd; SP-D
UniProt ID: (Mouse) P50404
Entrez Gene ID: (Mouse) 20390
Molecular Function: defense/immunity protein surfactant
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.