Invitrogen
Manufacturer Code:PA542014
Catalog # PIPA542014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SFTPC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRICHOS domain containing 6; pulmonary surfactant apoprotein-2 SP-C; pulmonary surfactant associated protein C; Pulmonary surfactant-associated protein C; Pulmonary surfactant-associated proteolipid SPL(Val); SP-C; SP5; surfactant associated protein C; Surfactant protein C PSP-C SFTP2 SMDP2 SP-C; Surfactant pulmonary-associated protein C; Surfactant, pulmonary-associated protein C; surfactant-associated protein; type I SP-C; type II SP-C
Gene Aliases: BRICD6; pro-SpC; PSP-C; Sftp-2; SFTP2; SFTPC; SMDP2; SP-C; SP5; SPC
UniProt ID: (Rabbit) P22398, (Human) Q7Z5D0, (Rat) P11685
Entrez Gene ID: (Dog) 477385, (Sheep) 443034, (Rabbit) 100009162, (Horse) 100053773, (Human) 6440, (Mouse) 20389, (Rat) 50683, (Guinea pig) 100728764
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.