Invitrogen
Manufacturer Code:MA513975
Catalog # PIMA513975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SPB01 (1-2-A5-9) |
Immunogen | Recombit pro-surfactant protein B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa pulmonary-surfactant protein; 18 kDa pulmonary-surfactant protein 6 kDa proteinpulmonary surfactant-associated protein B pulmonary surfactant-associated proteolipid SPL(Phe); 6 kDa protein; Pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SP-B; surfactant associated protein B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B
Gene Aliases: PSP-B; SFTB3; SFTP3; SFTPB; SMDP1; SP-B
UniProt ID: (Human) P07988, (Rat) P22355
Entrez Gene ID: (Human) 6439, (Rat) 192155
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.