Invitrogen
Manufacturer Code:PA583246
Catalog # PIPA583246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SFTPB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa pulmonary-surfactant protein; 6 kDa protein; Pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SFTP3 SP-B; SP-B
Gene Aliases: PSP-B; SFTB3; SFTP3; SFTPB; SMDP1; SP-B
UniProt ID: (Human) P07988
Entrez Gene ID: (Human) 6439
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.