Invitrogen
Manufacturer Code:PA552679
Catalog # PIPA552679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SFRP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Frizzled protein, human endometrium; FRP-4 FRPHE frizzled protein human endometrium sFRP-4 secreted frizzled-related protein 4 secreted frizzled-related protein 4; FrpHE; Secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; sFRP-4
Gene Aliases: FRP-4; FRPHE; sFRP-4; SFRP4
UniProt ID: (Human) Q6FHJ7
Entrez Gene ID: (Human) 6424
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.