Invitrogen
Manufacturer Code:MA526867
Catalog # PIMA526867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6E1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 25-295 of SFRP2 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Frizzled related protein 2 FRP 2 FRP-2 FRP2 MGC128911 MGC153618 MGC53344 SARP 1 SARP-1 SARP1 SDF 5 SDF5 Secreted apoptosis related protein 1 Secreted apoptosis-related protein 1 Secreted frizzled related protein 2 Secreted frizzled-related protein 2 sFRP 2 sFRP-2 SFRP2 Sfrp2 protein; FRP-2; SARP-1; secreted apoptosis related protein 1; Secreted apoptosis-related protein 1; Secreted frizzled-related protein 2; sFRP-2; testicular tissue protein Li 170
Gene Aliases: FKSG12; FRP-2; FRP2; SARP1; SDF-5; SFRP2; UNQ361/PRO697
UniProt ID: (Human) Q96HF1
Entrez Gene ID: (Human) 6423
Molecular Function: G-protein coupled receptor receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.