Invitrogen
Manufacturer Code:PA576794
Catalog # PIPA576794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SFRP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Frizzled related protein 2 FRP 2 FRP-2 FRP2 MGC128911 MGC153618 MGC53344 SARP 1 SARP-1 SARP1 SDF 5 SDF5 Secreted apoptosis related protein 1 Secreted apoptosis-related protein 1 Secreted frizzled related protein 2 Secreted frizzled-related protein 2 sFRP 2 sFRP-2 SFRP2 Sfrp2 protein; FRP-2; Protein SDF5; SARP-1; secreted apoptosis related protein 1; Secreted apoptosis-related protein 1; Secreted frizzled-related protein 2; secreted frizzled-related sequence protein 2; secreted frizzled-related sequence protein 5; sFRP-2; stromal cell derived factor 5; testicular tissue protein Li 170
Gene Aliases: AI851596; FKSG12; FRP-2; FRP2; SARP1; SDF-5; Sdf5; SFRP2; UNQ361/PRO697
UniProt ID: (Human) Q96HF1, (Mouse) P97299
Entrez Gene ID: (Human) 6423, (Rat) 310552, (Mouse) 20319
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.