Invitrogen
Manufacturer Code:MA538193
Catalog # PIMA538193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1683 |
Immunogen | A synthesized peptide derived from human SFRP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2210415K03Rik AW011917 AW107218 AW742929 frizzled-related protein FRP FRP1 FRP-1 FrzA SARP2 SARP-2 Secreted apoptosis-related protein 2 secreted frizzled related protein 1 secreted frizzled-related protein 1 secreted frizzled-related sequence protein 1 Sfrp1 sFRP-1; frizzled-related protein; FRP-1; SARP-2; Secreted apoptosis-related protein 2; Secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; sFRP-1
Gene Aliases: 2210415K03Rik; AW011917; AW107218; AW742929; FRP; FRP-1; FRP1; FrzA; SARP2; sFRP-1; SFRP1
UniProt ID: (Human) Q8N474, (Mouse) Q8C4U3
Entrez Gene ID: (Human) 6422, (Mouse) 20377, (Rat) 84402
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.