Invitrogen
Manufacturer Code:PA519004
Catalog # PIPA519004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KNHECPTFQSVFK) corresponding to the C-terminus amino acids of SFRP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: frizzled-related protein; FRP-1; SARP-2; Secreted apoptosis-related protein 2; Secreted frizzled-related protein 1; sFRP-1; SFRP1 secreted frizzled-related protein 1 FRP FRP-1 FRP1 FrzA SARP2 secreted apoptosis-related protein 2
Gene Aliases: FRP; FRP-1; FRP1; FrzA; SARP2; SFRP1
UniProt ID: (Human) Q8N474
Entrez Gene ID: (Human) 6422
Molecular Function:
G-protein coupled receptor
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.