Invitrogen
Manufacturer Code:PA529437
Catalog # PIPA529437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 357 and 611 of Human SFPQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa DNA-pairing protein; DNA-binding p52/p100 complex, 100 kDa subunit; hPOMp100; polypyrimidine tract binding protein associated; polypyrimidine tract-binding protein-associated splicing factor; Polypyrimidine tract-binding protein-associated-splicing factor; POMP100 PSF 100 kDa DNA-pairing protein DNA-binding p52/p100 complex 100 kDa subunit PTB-associated splicing factor PTB-associated-splicing factor hPOMp100 polypyrimidine tract binding protein associated polypyrimidine tract-binding protein-associated splicing facto; protein phosphatase 1, regulatory subunit 140; PSF; PTB-associated splicing factor; PTB-associated-splicing factor; splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated); splicing factor proline/glutamine rich (polypyrimidine tract-binding protein-associated); splicing factor proline/glutamine-rich; Splicing factor, proline- and glutamine-rich
Gene Aliases: POMP100; PPP1R140; PSF; SFPQ
UniProt ID: (Human) P23246
Entrez Gene ID: (Human) 6421
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.