Invitrogen
Manufacturer Code:PA5100111
Catalog # PIPA5100111
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SFPQ corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa DNA-pairing protein; 100 kDa DNA-pairing protein 1110004P21Rik 2810416M14Rik 5730453G22Rik 9030402K04Rik AU021830 D4Ertd314e DNA-binding p52/p100 complex 100 kDa subunit Gm12940 hPOMp100 NonO/p54nrb homolog OTTMUSG00000009329 polypyrimidine tract binding protein associated polypyrimidine tract-binding protein-associated splicing factor Polypyrimidine tract-binding protein-associated-splicing factor POMP100 PPP1R140 protein phosphatase 1 regulatory subunit 140 PSF PTB-associated splicing factor PTB-associated-splicing factor REP1 Sfpq splicing factor proline and glutamine rich splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated) splicing factor proline/glutamine rich (polypyrimidine tract-binding protein-associated) splicing factor proline/glutamine-rich splicing factor proline- and glutamine-rich; DNA-binding p52/p100 complex, 100 kDa subunit; hPOMp100; NonO/p54nrb homolog; polypyrimidine tract binding protein associated; polypyrimidine tract-binding protein-associated splicing factor; Polypyrimidine tract-binding protein-associated-splicing factor; protein phosphatase 1, regulatory subunit 140; PSF; PTB-associated splicing factor; PTB-associated-splicing factor; splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated); splicing factor proline/glutamine rich (polypyrimidine tract-binding protein-associated); splicing factor proline/glutamine-rich; Splicing factor, proline- and glutamine-rich
Gene Aliases: 1110004P21Rik; 2810416M14Rik; 5730453G22Rik; 9030402K04Rik; AU021830; D4Ertd314e; Gm12940; OTTMUSG00000009329; POMP100; PPP1R140; PSF; REP1; SFPQ
UniProt ID: (Human) P23246, (Mouse) Q8VIJ6
Entrez Gene ID: (Human) 6421, (Mouse) 71514, (Rat) 252855
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.