Novus Biologicals
Manufacturer Code:NBP157303
Catalog # NBP157303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SFPQ(splicing factor proline/glutamine-rich (polypyrimidine tract binding protein associated)) The peptide sequence was selected from the middle region of SFPQ. Peptide sequence PVIVEPLEQLDDEDGLPEKLAQKNP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa DNA-pairing protein; 100 kDa DNA-pairing protein DNA-binding p52/p100 complex 100 kDa subunit hPOMp100 polypyrimidine tract binding protein associated polypyrimidine tract-binding protein-associated splicing factor Polypyrimidine tract-binding protein-associated-splicing factor PSFPOMP100 PTB-associated splicing factor PTB-associated-splicing factor splicing factor proline/glutamine rich (polypyrimidine tract binding proteinassociated) splicing factor proline/glutamine rich (polypyrimidine tract-bindingprotein-associated) splicing factor proline/glutamine-rich splicing factor proline- and glutamine-rich; DNA-binding p52/p100 complex, 100 kDa subunit; hPOMp100; polypyrimidine tract binding protein associated; polypyrimidine tract-binding protein-associated splicing factor; Polypyrimidine tract-binding protein-associated-splicing factor; protein phosphatase 1, regulatory subunit 140; PSF; PTB-associated splicing factor; PTB-associated-splicing factor; splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated); splicing factor proline/glutamine rich (polypyrimidine tract-binding protein-associated); splicing factor proline/glutamine-rich; Splicing factor, proline- and glutamine-rich
Gene Aliases: POMP100; PPP1R140; PSF; SFPQ
UniProt ID: (Human) P23246
Entrez Gene ID: (Human) 6421
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.