Invitrogen
Manufacturer Code:PA561969
Catalog # PIPA561969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SF3A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: pre-mRNA splicing factor SF3A, subunit 2; PRP11 PRPF11 SAP62 SF3a66 SAP 62 pre-mRNA splicing factor SF3A subunit 2 spliceosome associated protein 62 spliceosome-associated protein 62 splicing factor 3A subunit 2; SAP 62; SF3a66; spliceosome associated protein 62; Spliceosome-associated protein 62; Splicing factor 3A subunit 2; splicing factor 3a, subunit 2, 66kD; splicing factor 3a, subunit 2, 66kDa
Gene Aliases: PRP11; PRPF11; SAP62; SF3A2; SF3a66
UniProt ID: (Human) Q15428
Entrez Gene ID: (Human) 8175
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.