Novus Biologicals
Manufacturer Code:NBP274150AF532
Catalog # NP27410A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F4 |
Immunogen | Recombinant protein fragment corresponding to amino acids 249-568 of human SF3A1 (NP_005868) produced in E.coli. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: pre-mRNA processing 21; pre-mRNA processing 21 pre-mRNA splicing factor SF3a (120 kDa subunit) Prp21 PRPF21 SAP 114 SAP114splicing factor 3a subunit 1 120kD SF3a120 spliceosome associated protein 114 Spliceosome-associated protein 114 splicing factor 3 subunit 1 splicing factor 3A subunit 1 splicing factor 3a subunit 1 120kDa; pre-mRNA splicing factor SF3a (120 kDa subunit); SAP 114; SF3a120; Spliceosome-associated protein 114; splicing factor 3 subunit 1; Splicing factor 3A subunit 1; splicing factor 3a, subunit 1, 120kD; splicing factor 3a, subunit 1, 120kDa
Gene Aliases: PRP21; PRPF21; SAP114; SF3A1; SF3A120
UniProt ID: (Human) Q15459
Entrez Gene ID: (Human) 10291
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.