Invitrogen
Manufacturer Code:PA541967
Catalog # PIPA541967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BBP; Mammalian branch point-binding protein; Splicing factor 1; Splicing factor 1 BBP MBBP ZFM1 ZNF162 D11S636; Transcription factor ZFM1; Zinc finger gene in MEN1 locus; Zinc finger protein 162
Gene Aliases: BBP; D11S636; MBBP; MZFM; SF1; WBP4; wu:fc09f06; ZCCHC25; ZFM1; Zfp162; ZNF162
UniProt ID: (Human) Q15637
Entrez Gene ID: (Human) 7536, (Rabbit) 100328702, (Dog) 476031, (Bovine) 541229, (Horse) 100050420, (Guinea pig) 100735823, (Mouse) 22668, (Rat) 117855, (Zebrafish) 572785
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.