Invitrogen
Manufacturer Code:PA528054
Catalog # PIPA528054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 309 of Human splicing factor 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BBP; BBP D11S636 MBBP ZFM1 ZNF162 mammalian branch point-binding protein transcription factor ZFM1 zinc finger gene in MEN1 locus zinc finger protein 162; Mammalian branch point-binding protein; Splicing factor 1; Transcription factor ZFM1; Zinc finger gene in MEN1 locus; Zinc finger protein 162
Gene Aliases: BBP; D11S636; MBBP; SF1; ZCCHC25; ZFM1; ZNF162
UniProt ID: (Human) Q15637
Entrez Gene ID: (Human) 7536
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.