Invitrogen
Manufacturer Code:PA518116
Catalog # PIPA518116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RWQKCVDCNGSYFD) corresponding to the C-terminus amino acids of SETMAR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Metnase; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion protein; SETMAR SET domain and mariner transposase fusion gene METNASE SET domain and mariner transposase fusion; Transposon Hsmar1 transposase
Gene Aliases: Mar1; METNASE; SETMAR
UniProt ID: (Human) Q53H47
Entrez Gene ID: (Human) 6419
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.