Invitrogen
Manufacturer Code:720343
Catalog # PI720343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Protein corresponding to human SETMAR &lsqbaa15-aa303] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epidermal tumor expressed transcript 2; Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Metnase; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion gene-containing protein homolog; SET domain and mariner transposase fusion protein; SET domain without mariner transposase fusion protein; SETMAR; Transposon Hsmar1 transposase
Gene Aliases: 5830404F24Rik; Etet2; Mar1; METNASE; SETMAR
UniProt ID: (Human) Q53H47, (Mouse) Q80UJ9
Entrez Gene ID: (Human) 6419, (Mouse) 74729
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.