Novus Biologicals
Manufacturer Code:NBP197740
Catalog # NBP197740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 225 and 275 of human SET Domain and Mariner Transposase Fusion Gene-Containing Protein using the numbering given in entry BAD96454.1 (GeneID 6419). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.43 histone-lysine N-methyltransferase SETMAR hsMar1 metnase SET domain and mariner transposase fusion gene SET domain and mariner transposase fusion gene-containing protein; Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Metnase; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion protein; Transposon Hsmar1 transposase
Gene Aliases: Mar1; METNASE; SETMAR
UniProt ID: (Human) Q53H47
Entrez Gene ID: (Human) 6419
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.