Novus Biologicals
Manufacturer Code:NB1001142
Catalog # NB1001142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence RWQKCVDCNGSYFD corresponding to C-Terminus according to NP_006506.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.43 histone-lysine N-methyltransferase SETMAR hsMar1 metnase SET domain and mariner transposase fusion gene SET domain and mariner transposase fusion gene-containing protein; Histone-lysine N-methyltransferase; Histone-lysine N-methyltransferase SETMAR; Metnase; SET domain and mariner transposase fusion gene-containing protein; SET domain and mariner transposase fusion protein; Transposon Hsmar1 transposase
Gene Aliases: Mar1; METNASE; SETMAR
UniProt ID: (Human) Q53H47
Entrez Gene ID: (Human) 6419
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.