Invitrogen
Manufacturer Code:PA518112
Catalog # PIPA518112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (GEKNGDAKTFWME) corresponding to the N-terminus amino acids of SETDB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Chronic lymphocytic leukemia deletion region gene 8 protein; Histone-lysine N-methyltransferase SETDB2; Lysine N-methyltransferase 1F; SET domain bifurcated 2; SETDB2 CLLD8 SET domain bifurcated 2 CLLL8 protein C13orf4 CLLL8 DKFZp586I0123 DKFZp761J1217 KMT1F chronic lymphocytic leukemia deletion region 8 lysine N-methyltransferase 1F histone-lysine N-methyltransferase SETDB2
Gene Aliases: C13orf4; CLLD8; CLLL8; KMT1F; SETDB2
UniProt ID: (Human) Q96T68
Entrez Gene ID: (Human) 83852
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.