Invitrogen
Manufacturer Code:PA5111677
Catalog # PIPA5111677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human SETDB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bifurcated 2 C13orf4 Chronic lymphocytic leukemia deletion region gene 8 protein CLLD8 CLLL8 CLLL8 protein DKFZp586I0123 DKFZp761J1217 Gm293 Histone-lysine N-methyltransferase SETDB2 KMT1F Lysine N-methyltransferase 1F SET domain SET domain bifurcated 2 SETDB2; Chronic lymphocytic leukemia deletion region gene 8 protein; Histone-lysine N-methyltransferase SETDB2; Lysine N-methyltransferase 1F; SET domain bifurcated 2
Gene Aliases: C13orf4; CLLD8; CLLL8; KMT1F; SETDB2
UniProt ID: (Human) Q96T68
Entrez Gene ID: (Human) 83852
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.