Novus Biologicals
Manufacturer Code:NBP220323
Catalog # NBP220323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human SETDB2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C13orf4 chromosome 13 open reading frame 4 chronic lymphocytic leukemia deletion region 8 Chronic lymphocytic leukemia deletion region gene 8 protein CLLD8DKFZp761J1217 DKFZp586I0123 EC 2.1.1.43 histone-lysine N-methyltransferase SETDB2 KMT1FCLLL8 Lysine N-methyltransferase 1F SET domain bifurcated 2 SET domain bifurcated 2; Chronic lymphocytic leukemia deletion region gene 8 protein; Histone-lysine N-methyltransferase SETDB2; Lysine N-methyltransferase 1F; SET domain bifurcated 2
Gene Aliases: C13orf4; CLLD8; CLLL8; KMT1F; SETDB2
UniProt ID: (Human) Q96T68
Entrez Gene ID: (Human) 83852
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.