Abnova
Manufacturer Code:PAB5573
Catalog # 89-131-912
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human SETDB2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SETDB2.
For Research Use Only
Protein Aliases: C13orf4; Chronic lymphocytic leukemia deletion region gene 8 protein; CLLD8; CLLL8; DKFZp586I0123; DKFZp761J1217; Histone-lysine N-methyltransferase SETDB2; KMT1F; Lysine N-methyltransferase 1F; SET domain bifurcated 2
Gene Aliases: C13orf4; CLLD8; CLLL8; KMT1F; SETDB2
UniProt ID: (Human) Q96T68
Entrez Gene ID: (Human) 83852
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.