Proteintech
Manufacturer Code:112311AP150UL
Catalog # 50-555-080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SETDB1 Fusion Protein Ag1725 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERG-associated protein with a SET domain, ESET; ERG-associated protein with SET domain; ESET; ESET H3 K9 HMTase 4 H3 K9 HMTase4 KG1T KIAA0067 KMT1E Lysine N methyltransferase 1E SET domain bifurcated 1 SET domain bifurcated 1 SETDB1; H3-K9-HMTase 4; Histone H3-K9 methyltransferase 4; Histone-lysine N-methyltransferase SETDB1; histone-lysine N-methyltransferase, H3lysine-9 specific 4; Lysine N-methyltransferase 1E; SET domain bifurcated 1; SET domain ERG-associated histone methyltransferase; SET domain, bifurcated 1; tudor domain containing 21
Gene Aliases: AU022152; ESET; H3-K9-HMTase4; KG1T; KIAA0067; KMT1E; mKIAA0067; SETDB1; TDRD21
UniProt ID: (Human) Q15047
Entrez Gene ID: (Human) 9869, (Mouse) 84505, (Rat) 689883
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.