Novus Biologicals
Manufacturer Code:NBP220320
Catalog # NBP220320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human SETD8. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: H4-K20-HMTase KMT5A; H4-K20-HMTase SETD8; H4-K20-HMTase SETD8 H4K20-specific histone methyltransferase splice variant Set8b Histone-lysine N-methyltransferase SETD8 KMT5A KMT5A SET domain-containing protein 8 Lysine N-methyltransferase 5A PR/SET domain containing protein 8 PR/SET domain-containing protein 07 PR/SET07 PRSET7 PR-Set7EC 2.1.1.43 SET domain containing (lysine methyltransferase) 8 SET07SET8H4-K20-specific histone methyltransferase SET8; H4-K20-specific histone methyltransferase; H4K20-specific histone methyltransferase splice variant Set8b; Histone-lysine N-methyltransferase KMT5A; histone-lysine N-methyltransferase SETD8; lysine (K)-specific methyltransferase 5A; Lysine N-methyltransferase 5A; Lysine-specific methylase 5A; N-lysine methyltransferase KMT5A; N-lysine methyltransferase SETD8; PR-Set7; PR/SET domain containing protein 8; PR/SET domain-containing protein 07; PR/SET07; SET domain containing (lysine methyltransferase) 8; SET domain-containing protein 8
Gene Aliases: KMT5A; PR-Set7; PRSET7; SET07; SET8; SETD8
UniProt ID: (Human) Q9NQR1
Entrez Gene ID: (Human) 387893
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.