Invitrogen
Manufacturer Code:PA511282
Catalog # PIPA511282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 159-189 amino acids from the central region of human SET7 (SET9) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H3-K4 methyltransferase; H3-K4-HMTase SETD7; Histone H3-K4 methyltransferase SETD7; histone H3-lysine 4-specific methyltransferase; Histone-lysine N-methyltransferase SETD7; KMT7; Lysine N-methyltransferase 7; SET domain containing (lysine methyltransferase) 7; SET domain-containing protein 7; SET7/9
Gene Aliases: 1600028F23Rik; H3K4MT; KIAA1717; KMT7; mKIAA1717; SET7; SET7/9; SET9; SETD7
UniProt ID: (Human) Q8WTS6, (Mouse) Q8VHL1
Entrez Gene ID: (Human) 80854, (Mouse) 73251
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.