Novus Biologicals
Manufacturer Code:NBP180046
Catalog # NBP180046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal of human C21ORF18. Peptide sequence LTALKLLCLEAEKFTCWKKVLLGEVISDTNEKTSLDIAQKICYYFIEETN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C21orf18 C21orf27 chromosome 21 open reading frame 18 chromosome 21 open reading frame 27 SET domain containing 4 SET domain-containing protein 4; SET domain-containing protein 4
Gene Aliases: C21orf18; C21orf27; SETD4
UniProt ID: (Human) Q9NVD3
Entrez Gene ID: (Human) 54093
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.