Invitrogen
Manufacturer Code:PA582105
Catalog # PIPA582105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SETD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-histidine N-methyltransferase; C14orf154 FLJ23027; Endothelial differentiation inhibitory protein D10; histone-lysine N-methyltransferase setd3; hSETD3; SET domain-containing protein 3
Gene Aliases: 2610102I01Rik; 2610305M23Rik; 6030490B17Rik; C14orf154; D12Ertd771e; RGD1309550; SETD3
UniProt ID: (Human) Q86TU7, (Rat) G3V6U9, (Mouse) Q91WC0
Entrez Gene ID: (Human) 84193, (Rat) 299295, (Mouse) 52690
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.