Abnova
Manufacturer Code:PAB15851
Catalog # 89-117-913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 254 amino acids of mouse Setd2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Setd2.
For Research Use Only
Protein Aliases: 4921524K10Rik; BC031601; Histone-lysine N-methyltransferase SETD2; KMT3A; Lysine N-methyltransferase 3A; Protein-lysine N-methyltransferase SETD2; SET domain-containing protein 2
Gene Aliases: 4921524K10Rik; BC031601; Kiaa1732; KMT3A; Setd2
UniProt ID: (Mouse) E9Q5F9
Entrez Gene ID: (Mouse) 235626
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.