OriGene
Manufacturer Code:CF804268
Catalog # 50-166-6911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1787-2144 of human SETD2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HBP231 HIF-1 HIP-1 HSPC069 HYPB KMT3A LLS SET2 p231HBP; HIF-1; HIP-1; Histone-lysine N-methyltransferase SETD2; hSET2; huntingtin interacting protein 1; Huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; Lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain-containing protein 2
Gene Aliases: HBP231; HIF-1; HIF1; HIP-1; HSPC069; HYPB; KIAA1732; KMT3A; LLS; p231HBP; SET2; SETD2
UniProt ID: (Human) Q9BYW2
Entrez Gene ID: (Human) 29072
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.