Invitrogen
Manufacturer Code:PA588104
Catalog # PIPA588104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 803-1103 of human SETD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Histone-lysine N-methyltransferase SETD2; kinesin family member 9; Lysine N-methyltransferase 3A; Protein-lysine N-methyltransferase SETD2; SET domain-containing protein 2
Gene Aliases: 4921524K10Rik; BC031601; Kiaa1732; Kif9; KMT3A; Setd2
UniProt ID: (Mouse) E9Q5F9
Entrez Gene ID: (Rat) 316013, (Mouse) 235626
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.