Invitrogen
Manufacturer Code:PA543071
Catalog # PIPA543071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SETD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HIF-1; HIP-1; Histone-lysine N-methyltransferase SETD2; hSET2; huntingtin interacting protein 1; Huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; kinesin family member 9; Lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain containing 2 FLJ16420 FLJ22472 FLJ23184 FLJ45883 HIF-1 HSPC069 HYPB KIAA1732 KMT3A SET2 HIP-1 HBP231 p231HBP; SET domain-containing protein 2
Gene Aliases: 4921524K10Rik; BC031601; HBP231; HIF-1; HIF1; HIP-1; HSPC069; HYPB; KIAA1732; Kif9; KMT3A; LLS; p231HBP; SET2; SETD2
UniProt ID: (Human) Q9BYW2, (Mouse) E9Q5F9
Entrez Gene ID: (Human) 29072, (Dog) 476643, (Horse) 100064888, (Rat) 316013, (Mouse) 235626
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.