Invitrogen
Manufacturer Code:PA5102710
Catalog # PIPA5102710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SETD2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4921524K10Rik BC031601 FLJ16420 FLJ22472 FLJ23184 FLJ45883 HBP231 HIF1 HIF-1 HIP-1 Histone-lysine N-methyltransferase SETD2 hSET2 HSPC069 huntingtin interacting protein 1 huntingtin yeast partner B Huntingtin-interacting protein 1 Huntingtin-interacting protein B HYPB Kiaa1732 Kif9 kinesin family member 9 KMT3A LLS Lysine N-methyltransferase 3A p231HBP Protein-lysine N-methyltransferase SETD2 SET domain containing 2 SET domain-containing protein 2 SET2 SETD2; HIF-1; HIP-1; Histone-lysine N-methyltransferase SETD2; hSET2; huntingtin interacting protein 1; Huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; kinesin family member 9; Lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain-containing protein 2
Gene Aliases: 4921524K10Rik; BC031601; HBP231; HIF-1; HIF1; HIP-1; HSPC069; HYPB; KIAA1732; Kif9; KMT3A; LLS; p231HBP; SET2; SETD2
UniProt ID: (Human) Q9BYW2, (Mouse) E9Q5F9
Entrez Gene ID: (Human) 29072, (Rat) 316013, (Mouse) 235626
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.