Abnova
Manufacturer Code:PAB23943
Catalog # 89-125-649
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SETD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SETD2.
For Research Use Only
Protein Aliases: FLJ16420; FLJ22472; FLJ23184; FLJ45883; FLJ46217; HIF-1; HIP-1; Histone-lysine N-methyltransferase SETD2; hSET2; HSPC069; huntingtin interacting protein 1; Huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; HYPB; KIAA1732; KMT3A; Lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain-containing protein 2; SET2
Gene Aliases: HBP231; HIF-1; HIF1; HIP-1; HSPC069; HYPB; KIAA1732; KMT3A; LLS; p231HBP; SET2; SETD2
UniProt ID: (Human) Q9BYW2
Entrez Gene ID: (Human) 29072
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.