Invitrogen
Manufacturer Code:PA5116923
Catalog # PIPA5116923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human SETD1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA516740 BC035291 histone-lysine N-methyltransferase SETD1B hSET1B Kiaa1076 KMT2G Lysine N-methyltransferase 2G mKIAA1076 SET domain containing 1B SET domain-containing protein 1B Set1b Setd1b; Histone-lysine N-methyltransferase SETD1B; hSET1B; Lysine N-methyltransferase 2G; rCG21620-like; SET domain-containing protein 1B
Gene Aliases: AA516740; BC035291; KIAA1076; KMT2G; mKIAA1076; SET1B; SETD1B
UniProt ID: (Human) Q9UPS6, (Mouse) Q8CFT2
Entrez Gene ID: (Rat) 100359816, (Human) 23067, (Mouse) 208043
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.