Abnova
Manufacturer Code:PAB21443
Catalog # 89-122-938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SETD1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SETD1B.
For Research Use Only
Protein Aliases: FLJ20803; Histone-lysine N-methyltransferase SETD1B; hSET1B; KIAA1076; KMT2G; Lysine N-methyltransferase 2G; SET domain-containing protein 1B; Set1B
Gene Aliases: KIAA1076; KMT2G; SET1B; SETD1B
UniProt ID: (Human) Q9UPS6
Entrez Gene ID: (Human) 23067
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.