Abnova
Manufacturer Code:PAB14084
Catalog # 89-116-920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A mixture of synthetic peptides (conjugated with KLH) that corresponding to internal region of mouse Setd1b. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Setd1b.
For Research Use Only
Protein Aliases: AA516740; BC035291; Histone-lysine N-methyltransferase SETD1B; KMT2G; MGC39035; mKIAA1076; SET domain-containing protein 1B
Gene Aliases: AA516740; BC035291; Kiaa1076; KMT2G; mKIAA1076; Set1b; Setd1b
UniProt ID: (Mouse) Q8CFT2
Entrez Gene ID: (Mouse) 208043
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.